Tisk…

3-(Isopropylamino)propanol 95%

Dodavatel: Apollo Scientific

Synonyms: 3-(Isopropylamino)propan-1-ol

OR938476-25G OR938476-5G
APOSOR938476-25GEA 9100 CZK
APOSOR938476-25G APOSOR938476-5G
3-(Isopropylamino)propanol 95%
3-(Isopropylamino)propanol
Vzorec: C₆H₁₅NO
MW: 117,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00483086
CAS číslo: 33918-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR