Butane-2-sulphonyl chloride 95%
Dodavatel: Apollo Scientific
OR938350-1G
OR938350-5G
APOSOR938350-1GEA
2110
CZK
APOSOR938350-1G
APOSOR938350-5G
Butane-2-sulphonyl chloride 95%
Butane-2-sulphonyl chloride
Vzorec:
C₄H₉ClO₂S Storage Temperature: Ambient |
MDL Number:
MFCD04973899 CAS číslo: 4375-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...