AmbotzHAA6300 95%
Dodavatel: Apollo Scientific
Synonyms:
H-D-Leu-OtBu·HCl
OR938329-10G
OR938329-1G
OR938329-5G
APOSOR938329-10GEA
27480
CZK
APOSOR938329-10G
APOSOR938329-1G
APOSOR938329-5G
AmbotzHAA6300 95%
AmbotzHAA6300
Vzorec:
C₁₀H₂₁NO₂ MW: 187,283 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00083642 CAS číslo: 67617-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...