Tisk…

5-Bromoresorcinol 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Bromo-3,5-dihydroxybenzene, 5-Bromobenzene-1,3-diol, 3,5-Dihydroxybromobenzene

OR938303-5G
APOSOR938303-5GEA 2730 CZK
APOSOR938303-5G
5-Bromoresorcinol 97%
5-Bromoresorcinol
Vzorec: C₆H₅BrO₂
MW: 189,01 g/mol
Bod varu: 140 °C/0,3mmHg
Bod tání: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD16293393
CAS číslo: 106120-04-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR