Tisk…

2,2-Dimethylbutyryl chloride 97%

Dodavatel: Apollo Scientific

Synonyms: 2,2-Dimethylbutyrylchloride

OR938206-500G
APOSOR938206-500GEA 3310 CZK
APOSOR938206-500G
2,2-Dimethylbutyryl chloride 97%
2,2-Dimethylbutyryl chloride
Vzorec: C₆H₁₁ClO
MW: 134,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093071
CAS číslo: 5856-77-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR