Methyl-1-methylcyclopropane-1-carboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
1-Methylcyclopropane-1-carboxylicacidmethylester, 1-Methylcyclopropane-1-carboxylic Acid Methyl Ester
OR938181-1G
OR938181-5G
APOSOR938181-1GEA
4270
CZK
APOSOR938181-1G
APOSOR938181-5G
Methyl-1-methylcyclopropane-1-carboxylate 95%
Methyl-1-methylcyclopropane-1-carboxylate
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191549 CAS číslo: 6206-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...