3-Anilino-1-propanol 95%
Dodavatel: Apollo Scientific
Synonyms:
N-(3-Hydroxypropyl)aniline, 3-(Phenylamino)propan-1-ol
OR938169-5G
APOSOR938169-5GEA
5920
CZK
APOSOR938169-5G
3-Anilino-1-propanol 95%
3-Anilino-1-propanol
Vzorec:
C₉H₁₃NO MW: 151,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15206074 CAS číslo: 31121-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...