N-Methoxy-N-methylcarbamoyl chloride 95%
Dodavatel: Apollo Scientific
OR938165-1G
OR938165-5G
APOSOR938165-1GEA
2530
CZK
APOSOR938165-1G
APOSOR938165-5G
N-Methoxy-N-methylcarbamoyl chloride 95%
N-Methoxy-N-methylcarbamoyl chloride
Vzorec:
C₃H₆ClNO₂ MW: 123,54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658962 CAS číslo: 30289-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...