Tisk…
6-(1H-pyrazol-1-yl)nicotinic acid 97%
Dodavatel: Apollo Scientific
Synonyms: 6-(1H-Pyrazol-1-yl)nicotinic acid
OR938164-1G OR938164-5G
APOSOR938164-1GEA 2940 CZK
APOSOR938164-1G APOSOR938164-5G
6-(1H-pyrazol-1-yl)nicotinic acid 97%
6-(1H-pyrazol-1-yl)nicotinic acid
Vzorec: C₉H₇N₃O₂
MW: 189,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140749
CAS číslo: 253315-22-9