Tisk…

(R)-(-)-2-Methylglutaric acid 95%

Dodavatel: Apollo Scientific

OR938135-10G OR938135-1G OR938135-5G
APOSOR938135-10GEA 11150 CZK
APOSOR938135-10G APOSOR938135-1G APOSOR938135-5G
(R)-(-)-2-Methylglutaric acid 95%
(R)-(-)-2-Methylglutaric acid
Vzorec: C₆H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00671562
CAS číslo: 1115-81-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR