Tisk…

2-(2-Thienyl)-1,3-dioxolane 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(1,3-Dioxolan-2-yl)thiophene

OR938123-10G OR938123-1G OR938123-5G
APOSOR938123-10GEA 19440 CZK
APOSOR938123-10G APOSOR938123-1G APOSOR938123-5G
2-(2-Thienyl)-1,3-dioxolane 95%
2-(2-Thienyl)-1,3-dioxolane
Vzorec: C₇H₈O₂S
MW: 156.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01924576
CAS číslo: 58268-08-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR