Tisk…

Diisopropyl sulphite 95%

Dodavatel: Apollo Scientific

OR938058-25G OR938058-5G
APOSOR938058-25GEA 6420 CZK
APOSOR938058-25G APOSOR938058-5G
Diisopropyl sulphite 95%
Diisopropyl sulphite
Vzorec: C₆H₁₄O₃S
MW: 166.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200838
CAS číslo: 4773-13-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR