Ethyl-2-methyl-3-furancarboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Methyl-3-furancarboxylic Acid Ethyl Ester, 2-Methyl-3-furoic Acid Ethyl Ester, Ethyl 2-Methyl-3-furoate
OR938025-1G
OR938025-5G
APOSOR938025-1GEA
4140
CZK
APOSOR938025-1G
APOSOR938025-5G
Ethyl-2-methyl-3-furancarboxylate 95%
Ethyl-2-methyl-3-furancarboxylate
Vzorec:
C₈H₁₀O₃ MW: 154,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134327 CAS číslo: 28921-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...