2,5-Dimethylcyclohexanol (mixture of isomers) 95%
Dodavatel: Apollo Scientific
OR937948-25G
OR937948-5G
APOSOR937948-25GEA
8070
CZK
APOSOR937948-25G
APOSOR937948-5G
2,5-Dimethylcyclohexanol (mixture of isomers) 95%
2,5-Dimethylcyclohexanol (mixture of isomers)
Vzorec:
C₈H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD00001504 CAS číslo: 3809-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...