Allylpalladium(II) chloride dimer 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer
OR937920-1G
APOSOR937920-1GEA
4140
CZK
APOSOR937920-1G
Allylpalladium(II) chloride dimer 97%
Allylpalladium(II) chloride dimer
Vzorec:
C₆H₁₀Cl₂Pd₂ MW: 365,89 g/mol Bod tání: 120 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044874 CAS číslo: 12012-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...