Tisk…

4-Penten-1-yl isothiocyanate 95%

Dodavatel: Apollo Scientific

OR937905-1G OR937905-250MG
APOSOR937905-1GEA 9100 CZK
APOSOR937905-1G APOSOR937905-250MG
4-Penten-1-yl isothiocyanate 95%
4-Penten-1-yl isothiocyanate
Vzorec: C₆H₉NS
Storage Temperature: Ambient
MDL Number: MFCD00191527
CAS číslo: 18060-79-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR