Tisk…

2,2-dimethylbut-3-enoic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2,2-Dimethylbut-3-enoic acid

OR937784-1G OR937784-5G
APOSOR937784-1GEA 3060 CZK
APOSOR937784-1G APOSOR937784-5G
2,2-dimethylbut-3-enoic acid 95%
2,2-dimethylbut-3-enoic acid
Vzorec: C₆H₁₀O₂
MW: 114,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00196967
CAS číslo: 10276-09-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR