Tisk…

2-Chloro-N-ethylnicotinamide 95%

Dodavatel: Apollo Scientific

OR937765-1G OR937765-5G
APOSOR937765-1GEA 0 CZK
APOSOR937765-1G APOSOR937765-5G
2-Chloro-N-ethylnicotinamide 95%
2-Chloro-N-ethylnicotinamide
Vzorec: C₈H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD01358076
CAS číslo: 52943-22-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR