Tisk…

5-Norbornene-2,2-dimethanol 95%

Dodavatel: Apollo Scientific

OR937742-1G OR937742-5G
APOSOR937742-1GEA 2240 CZK
APOSOR937742-1G APOSOR937742-5G
5-Norbornene-2,2-dimethanol 95%
5-Norbornene-2,2-dimethanol
Vzorec: C₉H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00167595
CAS číslo: 6707-12-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR