Tisk…

Propionaldehyde dimethyl acetal 95%

Dodavatel: Apollo Scientific

OR937730-25G OR937730-1G
APOSOR937730-25GEA 0 CZK
APOSOR937730-25G APOSOR937730-1G
Propionaldehyde dimethyl acetal 95%
Propionaldehyde dimethyl acetal
Vzorec: C₅H₁₂O₂
MW: 104,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059307
CAS číslo: 4744-10-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR