Tisk…

Sulphur trioxide triethylamine complex 95%

Dodavatel: Apollo Scientific

OR937729-1G OR937729-25G OR937729-5G
APOSOR937729-1GEA 3540 CZK
APOSOR937729-1G APOSOR937729-25G APOSOR937729-5G
Sulphur trioxide triethylamine complex 95%
Sulphur trioxide triethylamine complex
Vzorec: C₆H₁₅NO₃S
Storage Temperature: Ambient
CAS číslo: 761-01-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR