Tisk…

Isoamyl acrylate 95%

Dodavatel: Apollo Scientific

Synonyms: Isopentyl Acrylate, Acrylicacidisoamylester, Acrylic Acid Isopentyl Ester, Acrylic Acid Isoamyl Ester

OR937646-25G OR937646-5G
APOSOR937646-25GEA 14290 CZK
APOSOR937646-25G APOSOR937646-5G
Isoamyl acrylate 95%
Isoamyl acrylate
Vzorec: C₈H₁₄O₂
MW: 142,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053725
CAS číslo: 4245-35-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR