3-(2-Nitroethenyl)pyridine 95%
Dodavatel: Apollo Scientific
Synonyms:
3-(2-Nitrovinyl)pyridine, 2-(3-Pyridyl)-1-nitroethylene
OR937645-10G
OR937645-1G
OR937645-5G
APOSOR937645-10GEA
27480
CZK
APOSOR937645-10G
APOSOR937645-1G
APOSOR937645-5G
3-(2-Nitroethenyl)pyridine 95%
3-(2-Nitroethenyl)pyridine
Vzorec:
C₇H₆N₂O₂ MW: 150,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191563 CAS číslo: 3156-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...