Tisk…

sec-Butyl-4-hydroxybenzoate 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Hydroxybenzoic acid sec-butyl ester

OR937629-100G OR937629-25G OR937629-5G
APOSOR937629-100GEA 19440 CZK
APOSOR937629-100G APOSOR937629-25G APOSOR937629-5G
sec-Butyl-4-hydroxybenzoate 95%
sec-Butyl-4-hydroxybenzoate
Vzorec: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00059626
CAS číslo: 17696-61-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR