3,3,5-Trimethylcyclohexylamine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR937606-1G
OR937606-5G
APOSOR937606-1GEA
1620
CZK
APOSOR937606-1G
APOSOR937606-5G
3,3,5-Trimethylcyclohexylamine 95%
3,3,5-Trimethylcyclohexylamine
Vzorec:
C₉H₁₉N MW: 141,26 g/mol Bod varu: 177 °C Hustota: 0,84 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00019379 CAS číslo: 15901-42-5 UN: 2326 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...