Heptaethylene glycol 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR937598-1G
OR937598-5G
APOSOR937598-1GEA
5630
CZK
APOSOR937598-1G
APOSOR937598-5G
Heptaethylene glycol 98%
Heptaethylene glycol
Vzorec:
C₁₄H₃₀O₈ Hustota: 1,13 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002876 CAS číslo: 5617-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...