3-phenylimidazolidine-2,4-dione 95%
Dodavatel: Apollo Scientific
Synonyms:
3-Phenylimidazolidine-2,4-dione, 3-Phenylhydantoin
OR937596-1G
OR937596-250MG
APOSOR937596-1GEA
4240
CZK
APOSOR937596-1G
APOSOR937596-250MG
3-phenylimidazolidine-2,4-dione 95%
3-phenylimidazolidine-2,4-dione
Vzorec:
C₉H₈N₂O₂ MW: 176.175 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209486 CAS číslo: 2221-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...