Tisk…

Trimethylsilylmethyl azide 95%

Dodavatel: Apollo Scientific

Synonyms: Azidomethyltrimethylsilane

OR937595-1G OR937595-250MG
APOSOR937595-1GEA 10150 CZK
APOSOR937595-1G APOSOR937595-250MG
Trimethylsilylmethyl azide 95%
Trimethylsilylmethyl azide
Vzorec: C₄H₁₁N₃Si
MW: 129,24 g/mol
Bod varu: 60 °C/86mmHg
Hustota: 0,87 g/cm³ (25 °C)
Bod vzplanutí: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00059964
CAS číslo: 87576-94-1
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR