Tisk…

Glycidyl stearate 90%

Dodavatel: Apollo Scientific

Synonyms: Stearic Acid Glycidyl Ester

OR937562-1G OR937562-25G OR937562-5G
APOSOR937562-1GEA 1870 CZK
APOSOR937562-1G APOSOR937562-25G APOSOR937562-5G
Glycidyl stearate 90%
Glycidyl stearate
Vzorec: C₂₁H₄₀O₃
MW: 340.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059110
CAS číslo: 7460-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR