Tisk…

Methyl (2S,3S)-2-isocyanato-3-methylvalerate 95%

Dodavatel: Apollo Scientific

Synonyms: (2S,3S)-2-Isocyanato-3-methylvaleric Acid Methyl Ester

OR937498-1G OR937498-5G
APOSOR937498-1GEA 5790 CZK
APOSOR937498-1G APOSOR937498-5G
Methyl (2S,3S)-2-isocyanato-3-methylvalerate 95%
Methyl (2S,3S)-2-isocyanato-3-methylvalerate

  • May form small amount of sediment.

Vzorec: C₈H₁₃NO₃
MW: 171,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191534
CAS číslo: 120219-17-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR