Tisk…

Tris(2-phenylpyridinato)iridium(III) 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Tris(2-phenylpyridine)iridium, Ir(ppy)3

OR937441-1G OR937441-250MG
APOSOR937441-1GEA 16530 CZK
APOSOR937441-1G APOSOR937441-250MG
Tris(2-phenylpyridinato)iridium(III) 95%
Tris(2-phenylpyridinato)iridium(III)
Vzorec: C₃₃H₂₄IrN₃
Storage Temperature: Ambient
MDL Number: MFCD04039766
CAS číslo: 94928-86-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR