Tisk…

(Tetrahydrofuran-2-yl)acetonitrile 95%

Dodavatel: Apollo Scientific

OR937427-1G OR937427-5G
APOSOR937427-1GEA 3710 CZK
APOSOR937427-1G APOSOR937427-5G
(Tetrahydrofuran-2-yl)acetonitrile 95%
(Tetrahydrofuran-2-yl)acetonitrile
Vzorec: C₆H₉NO
Storage Temperature: Ambient
MDL Number: MFCD03206782
CAS číslo: 33414-62-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR