2-Methoxyethyl chloroacetate 95%
Dodavatel: Apollo Scientific
Synonyms:
Chloroacetic acid 2-methoxyethyl ester
OR937425-10G
OR937425-1G
OR937425-5G
APOSOR937425-10GEA
14680
CZK
APOSOR937425-10G
APOSOR937425-1G
APOSOR937425-5G
2-Methoxyethyl chloroacetate 95%
2-Methoxyethyl chloroacetate
Vzorec:
C₅H₉ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00191626 CAS číslo: 13361-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...