N-Methyl-4-nitro-o-toluidine 95%
Dodavatel: Apollo Scientific
Synonyms:
N,2-Dimethyl-4-nitroaniline, 2-(Methylamino)-5-nitrotoluene
OR937419-1G
OR937419-5G
APOSOR937419-1GEA
7670
CZK
APOSOR937419-1G
APOSOR937419-5G
N-Methyl-4-nitro-o-toluidine 95%
N-Methyl-4-nitro-o-toluidine
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059083 CAS číslo: 10439-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...