4,4-Dimethyl-2-imidazolidinethione 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Mercapto-4,4-dimethylimidazoline, 4,4-Dimethylethylenethiourea
OR937418-5G
APOSOR937418-5GEA
5290
CZK
APOSOR937418-5G
4,4-Dimethyl-2-imidazolidinethione 95%
4,4-Dimethyl-2-imidazolidinethione
Vzorec:
C₅H₁₀N₂S MW: 130,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00661072 CAS číslo: 6086-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...