Tisk…

N-Methyl-3-nitroaniline 98%

Dodavatel: Apollo Scientific

OR937404-1G OR937404-5G
APOSOR937404-1GEA 3310 CZK
APOSOR937404-1G APOSOR937404-5G
N-Methyl-3-nitroaniline 98%
N-Methyl-3-nitroaniline
Vzorec: C₇H₈N₂O₂
MW: 152,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00963641
CAS číslo: 619-26-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR