Thionicotinic acid 95%
Dodavatel: Apollo Scientific
OR937397-10G
OR937397-1G
OR937397-5G
APOSOR937397-10GEA
0
CZK
APOSOR937397-10G
APOSOR937397-1G
APOSOR937397-5G
Thionicotinic acid 95%
Thionicotinic acid
Vzorec:
C₆H₅NOS Bod tání: 144…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191582 CAS číslo: 51087-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...