Tisk…

N-Chloroacetanilide 95%

Dodavatel: Apollo Scientific

Synonyms: N-Chloroacetanilide

OR937346-1G OR937346-250MG
APOSOR937346-1GEA 8070 CZK
APOSOR937346-1G APOSOR937346-250MG
N-Chloroacetanilide 95%
N-Chloroacetanilide
Vzorec: C₈H₈ClNO
MW: 169,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01671408
CAS číslo: 579-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR