Tisk…

Methyl-3-(4-chlorosulphonyl)phenylpropionate 97%

Dodavatel: Apollo Scientific

OR937304-25G OR937304-5G
APOSOR937304-25GEA 21820 CZK
APOSOR937304-25G APOSOR937304-5G
Methyl-3-(4-chlorosulphonyl)phenylpropionate 97%
Methyl-3-(4-chlorosulphonyl)phenylpropionate
Vzorec: C₁₀H₁₁ClO₄S
Bod varu: 352 °C
Bod tání: 64…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093942
CAS číslo: 374537-95-8
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR