Tisk…

2,2-Dimethyl-4-pentenoic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2,2-Dimethylpent-4-enoic acid

OR937287-5G
APOSOR937287-5GEA 3970 CZK
APOSOR937287-5G
2,2-Dimethyl-4-pentenoic acid 95%
2,2-Dimethyl-4-pentenoic acid
Vzorec: C₇H₁₂O₂
MW: 128,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012304
CAS číslo: 16386-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR