3,4-Dichlorothiophene-1,1-dioxide 95%
Dodavatel: Apollo Scientific
OR937277-250MG
APOSOR937277-250MGEA
25790
CZK
APOSOR937277-250MG
3,4-Dichlorothiophene-1,1-dioxide 95%
3,4-Dichlorothiophene-1,1-dioxide
Vzorec:
C₄H₂Cl₂O₂S MW: 185,02 g/mol Bod tání: 108 °C (rozkl.) Storage Temperature: Ambient |
CAS číslo:
52819-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...