Methylsuccinic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
Pyrotartaric Acid, 2-Methylsuccinic acid
OR937273-100G
APOSOR937273-100GEA
1910
CZK
APOSOR937273-100G
Methylsuccinic acid 95%
Methylsuccinic acid
Vzorec:
C₅H₈O₄ MW: 132,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002659 CAS číslo: 498-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...