1-(1,3-Thiazol-2-yl)propan-1-one 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Propionylthiazole, 1-(Thiazol-2-yl)propan-1-one
OR937249-25G
APOSOR937249-25GEA
2150
CZK
APOSOR937249-25G
1-(1,3-Thiazol-2-yl)propan-1-one 95%
1-(1,3-Thiazol-2-yl)propan-1-one
Vzorec:
C₆H₇NOS MW: 141,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01681404 CAS číslo: 43039-98-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...