2,5-Dimethoxy-1,4-benzoquinone 95%
Dodavatel: Apollo Scientific
Synonyms:
2,5-Dimethoxy-p-quinone
OR937216-1G
OR937216-5G
APOSOR937216-1GEA
827
CZK
APOSOR937216-1G
APOSOR937216-5G
2,5-Dimethoxy-1,4-benzoquinone 95%
2,5-Dimethoxy-1,4-benzoquinone
Vzorec:
C₈H₈O₄ MW: 168,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00189386 CAS číslo: 3117-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...