Tisk…

(1-Chloro-1-methylethyl)benzene 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9372-10G OR9372-25G OR9372-5G
APOSOR9372-10GEA 7050 CZK
APOSOR9372-10G APOSOR9372-25G APOSOR9372-5G
(1-Chloro-1-methylethyl)benzene 97%
(1-Chloro-1-methylethyl)benzene
Vzorec: H₃CCCl(C₆H₅)CH₃
MW: 154,64 g/mol
Bod varu: 48…50 °C/0,5mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00040715
CAS číslo: 934-53-2
EINECS: 213-284-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR