Tisk…

2-Methoxythiophene 95%

Dodavatel: Apollo Scientific

OR937091-25G
APOSOR937091-25GEA 2070 CZK
APOSOR937091-25G
2-Methoxythiophene 95%
2-Methoxythiophene
Vzorec: C₅H₆OS
MW: 114,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014526
CAS číslo: 16839-97-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR