3-(4-Methoxyphenyl)propionyl chloride 95%
Dodavatel: Apollo Scientific
Synonyms:
3-(4-Methoxyphenyl)propanoyl chloride
OR937079-1G
OR937079-5G
APOSOR937079-1GEA
1740
CZK
APOSOR937079-1G
APOSOR937079-5G
3-(4-Methoxyphenyl)propionyl chloride 95%
3-(4-Methoxyphenyl)propionyl chloride
Vzorec:
C₁₀H₁₁ClO₂ MW: 198,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02258671 CAS číslo: 15893-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...