Tisk…

3-Methylthiophene-2-carboxamide 98%

Dodavatel: Apollo Scientific

OR937055-1G OR937055-5G
APOSOR937055-1GEA 3540 CZK
APOSOR937055-1G APOSOR937055-5G
3-Methylthiophene-2-carboxamide 98%
3-Methylthiophene-2-carboxamide
Vzorec: C₆H₇NOS
MW: 141,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01208055
CAS číslo: 76655-99-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR