Tisk…

(S)-(+)-3-Methyl-1-pentanol 95%

Dodavatel: Apollo Scientific

OR937032-1G OR937032-250MG
APOSOR937032-1GEA 11380 CZK
APOSOR937032-1G APOSOR937032-250MG
(S)-(+)-3-Methyl-1-pentanol 95%
(S)-(+)-3-Methyl-1-pentanol
Vzorec: C₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00135167
CAS číslo: 42072-39-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR