Tisk…

Allyl phenyl carbonate 95%

Dodavatel: Apollo Scientific

Synonyms: Allyl Phenyl Carbonate, ['Carbonic acid Allyl Phenyl Ester']

OR937027-25G
APOSOR937027-25GEA 5690 CZK
APOSOR937027-25G
Allyl phenyl carbonate 95%
Allyl phenyl carbonate
Vzorec: C₁₀H₁₀O₃
MW: 178,1846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784342
CAS číslo: 16308-68-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR